MilliporeSigma
[Site-directed mutagenesis and sulfhydryl PEGylation of lysostaphin].
Sheng wu gong cheng xue bao = Chinese journal of biotechnology (2012-03-08)
Hong Wu, Wei Fang, Jing Yuan, Hui Peng, Xuecheng Zhang, Yongzhong Wang, Yazhong Xiao
ABSTRACT
The purpose of this paper is to establish sulfhydryl site-directed PEGylation method for lysostaphin and to evaluate effects of mutagenesis and modification of amino acid residue within putative linker on enzyme activity. On the basis of structural analysis of lysostaphin, amino acid 133-154 of tentative linker between the N-terminal and C-terminal domain were chosen as the candidate residues for site-directed mutagenesis to cysteine. Subsequently, sulfhydryl site-directed PEGylation was performed by reacting PEG-maleimide reagent with the newly introduced cysteine residue of the mutant lysostaphin. The Cys-mutant and PEG-modified proteins were both purified, and their enzymatic activity were further PEGylated lysostaphins. The mono-PEGylated lysostaphins were separated from unmodified lysostaphins through highly efficient one step method with Ni(2+)-NTA column chromatography. However, both Cys-mutant and PEGylated lysostaphin only retained partial activities of the wild-type enzyme. It suggests that sulfhydryl site-directed PEGylation modification of the tentative linker between the N-terminal and C-terminal domain may affect the catalytic activity of lysostaphin.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled