MilliporeSigma
  • Toxicity determined in vitro by morphological alterations and neutral red absorption.
Toxicity determined in vitro by morphological alterations and neutral red absorption.
Toxicology letters (1985-02-01)
E Borenfreund, J A Puerner
ABSTRACT
A method is described which combines the use of a visual morphological cytotoxicity assay with a quantitative neutral red (NR) spectrophotometric test, for the assessment of the effect of toxic agents on 3T3 cells in culture. These sensitive and reproducible assays lend themselves to a screening procedure of potential toxicants which can help reduce the use of animals for toxicity testing.
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Neutral Red based