MilliporeSigma
  • Rabankyrin-5 interacts with EHD1 and Vps26 to regulate endocytic trafficking and retromer function.
Rabankyrin-5 interacts with EHD1 and Vps26 to regulate endocytic trafficking and retromer function.
Traffic (Copenhagen, Denmark) (2012-01-31)
Jing Zhang, Calliste Reiling, James B Reinecke, Iztok Prislan, Luis A Marky, Paul L Sorgen, Naava Naslavsky, Steve Caplan
ABSTRACT
Rabankyrin-5 (Rank-5) has been implicated as an effector of the small GTPase Rab5 and plays an important role in macropinocytosis. We have now identified Rank-5 as an interaction partner for the recycling regulatory protein, Eps15 homology domain 1 (EHD1). We have demonstrated this interaction by glutathione S-transferase-pulldown, yeast two-hybrid assay, isothermal calorimetry and co-immunoprecipitation, and found that the binding occurs between the EH domain of EHD1 and the NPFED motif of Rank-5. Similar to EHD1, we found that Rank-5 colocalizes and interacts with components of the retromer complex such as vacuolar protein sorting 26 (Vps26), suggesting a role for Rank-5 in retromer-based transport. Indeed, depletion of Rank-5 causes mislocalization of Vps26 and affects both the retrieval of mannose 6-phosphate receptor transport to the Golgi from endosomes and biosynthetic transport. Moreover, Rank-5 is required for normal retromer distribution, as overexpression of a wild-type Rank-5-small interfering RNA-resistant construct rescues retromer mislocalization. Finally, we show that depletion of either Rank-5 or EHD1 impairs secretion of vesicular stomatitis virus glycoprotein. Overall, our data identify a new interaction between Rank-5 and EHD1, and novel endocytic regulatory roles that include retromer-based transport and secretion.
Sigma-Aldrich
Cathepsin G from human leukocytes, lyophilized powder, ≥60 units/mg protein (Bradford)