MilliporeSigma
  • Determining clinically important changes in range of motion in patients with Dupuytren's Contracture: secondary analysis of the randomized, double-blind, placebo-controlled CORD I study.
Determining clinically important changes in range of motion in patients with Dupuytren's Contracture: secondary analysis of the randomized, double-blind, placebo-controlled CORD I study.
Clinical drug investigation (2011-10-05)
Jörg Witthaut, Andrew G Bushmakin, Robert A Gerber, Joseph C Cappelleri, Marie-Pierre Hellio Le Graverand-Gastineau
ABSTRACT
Injectable collagenase Clostridium histolyticum is efficacious in correcting Dupuytren's contracture as assessed by changes in the angle of contracture and range of motion (ROM). However, clinically important changes in ROM have not been evaluated in depth. The objective of this secondary analysis of the CORD I trial was to identify severity levels using baseline ROM, estimate a clinically important difference (CID) for ROM, and link the results to collagenase treatment and patient satisfaction. In the CORD I trial, patients with Dupuytren's disease and joint contractures ≥20° were randomized to receive a maximum of three collagenase 0.58 mg or placebo injections into the cord of the affected hand at 30-day intervals. The primary endpoint was reduction in contracture to ≤5° 30 days after the last injection (day 30). The secondary endpoints, which are reported in this analysis, were ROM, physician- and patient-rated severity ('normal', 'mild', 'moderate', 'severe') and improvement, and treatment satisfaction. Linear regression was used to model data for severity classification and CID estimation for ROM based on physician and patient ratings. At baseline, mean ROM was 43.9° in the collagenase-treated joints (n = 197) and 45.3° in the placebo-treated joints (n = 102). On day 30, mean ROM was 80.7° in the collagenase-treated joints and 49.5° in the placebo-treated joints. The mean increase in ROM was 36.7° in the collagenase-treated joints (p < 0.001) and 4.0° in the placebo-treated joints (not significant). The estimated CID for ROM was 13.5° (95% CI 11.9, 15.1), reflecting a one-category change in severity. The mean increase in ROM exceeded the CID in the collagenase-treated joints but not in the placebo-treated joints; the difference between collagenase treatment and placebo in the mean increase in ROM also exceeded the CID, implying that the improvement with collagenase was clinically relevant. The severity classification for ROM was: ≥67.0° ('normal'), ≥54.3 and <67.0° ('mild'), ≥41.6 and <54.3° ('moderate'), and <41.6° ('severe'). More collagenase- than placebo-treated patients achieved 'normal' (81% vs 25%; p < 0.0001) status, and more collagenase- than placebo-treated patients reported being 'very/quite satisfied' (87% vs 32%; p < 0.001). Injectable collagenase significantly improves ROM and treatment satisfaction versus placebo. ROM improvements are clinically relevant as well as statistically significant. These data support the potential need to include ROM and physician- and patient-rated severity and satisfaction as standard assessments for Dupuytren's contracture treatment outcomes. ClinicalTrials.gov Identifier: NCT00528606; other study identification number: AUX-CC-857 (Auxilium Pharmaceuticals, Inc.).
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)