MilliporeSigma
  • A new nitrilase-producing strain named Rhodobacter sphaeroides LHS-305: biocatalytic characterization and substrate specificity.
A new nitrilase-producing strain named Rhodobacter sphaeroides LHS-305: biocatalytic characterization and substrate specificity.
Applied biochemistry and biotechnology (2011-09-23)
Chunsheng Yang, Xuedong Wang, Dongzhi Wei
ABSTRACT
The characteristics of the new nitrilase-producing strain Rhodobacter sphaeroides LHS-305 were investigated. By investigating several parameters influencing nitrilase production, the specific cell activity was ultimately increased from 24.5 to 75.0 μmol g(-1) min(-1), and hereinto, the choice of inducer proved the most important factor. The aromatic nitriles (such as 3-cyanopyridine and benzonitrile) were found to be the most favorable substrates of the nitrilase by analyzing the substrate spectrum. It was speculated that the unsaturated carbon atom attached to the cyano group was crucial for this type of nitrilase. The value of apparent K (m), substrate inhibition constant, and product inhibition constant of the nitrilase against 3-cyanopyridine were 4.5 × 10(-2), 29.2, and 8.6 × 10(-3) mol L(-1), respectively. When applied in nicotinic acid preparation, the nitrilase is able to hydrolyze 200 mmol L(-1) 3-cyanopyridine with 93% conversion rate in 13 h by 6.1 g L(-1) cells (dry cell weight).
Sigma-Aldrich
Nitrilase, recombinant, expressed in E. coli, ≥2.0 U/mg