MilliporeSigma
  • An acyloxymethyl ketone-based probe to monitor the activity of glutathionylspermidine amidase in Escherichia coli.
An acyloxymethyl ketone-based probe to monitor the activity of glutathionylspermidine amidase in Escherichia coli.
Chembiochem : a European journal of chemical biology (2011-09-03)
Cheong-Meng Chong, Shijay Gao, Bing-Yu Chiang, Wen-Hung Hsu, Ting-Chien Lin, Tzu-Chieh Chen, Chun-Hung Lin
ABSTRACT
Cellular redox conditions affect Gsp amidase activity in Escherichia coli. Guided by the structure and catalytic mechanism of the amidase, we designed and synthesized an acyloxymethyl ketone-based activity probe containing a biotin handle. This probe was used to monitor Gsp amidase activity in E. coli lysates that had been subjected to oxidative or methylglyoxal-induced stress.
Sigma-Aldrich
Amidase from Pseudomonas aeruginosa, recombinant, expressed in E. coli, buffered aqueous glycerol solution, hydroxamate transferase ≥200 units/mg protein (biuret)