MilliporeSigma
  • Klf1 affects DNase II-alpha expression in the central macrophage of a fetal liver erythroblastic island: a non-cell-autonomous role in definitive erythropoiesis.
Klf1 affects DNase II-alpha expression in the central macrophage of a fetal liver erythroblastic island: a non-cell-autonomous role in definitive erythropoiesis.
Molecular and cellular biology (2011-08-03)
Susanna Porcu, Maria F Manchinu, Maria F Marongiu, Valeria Sogos, Daniela Poddie, Isadora Asunis, Loredana Porcu, Maria G Marini, Paolo Moi, Antonio Cao, Frank Grosveld, Maria S Ristaldi
ABSTRACT
A key regulatory gene in definitive erythropoiesis is the erythroid Kruppel-like factor (Eklf or Klf1). Klf1 knockout (KO) mice die in utero due to severe anemia, while residual circulating red blood cells retain their nuclei. Dnase2a is another critical gene in definitive erythropoiesis. Dnase2a KO mice are also affected by severe anemia and die in utero. DNase II-alpha is expressed in the central macrophage of erythroblastic islands (CMEIs) of murine fetal liver. Its main role is to digest the DNA of the extruded nuclei of red blood cells during maturation. Circulating erythrocytes retain their nuclei in Dnase2a KO mice. Here, we show that Klf1 is expressed in CMEIs and that it binds and activates the promoter of Dnase2a. We further show that Dnase2a is severely downregulated in the Klf1 KO fetal liver. We propose that this downregulation of Dnase2a in the CMEI contributes to the Klf1 KO phenotype by a non-cell-autonomous mechanism.
Sigma-Aldrich
Deoxyribonuclease II from porcine spleen, Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein