MilliporeSigma
  • Reaction pathway of tryptophanase-catalyzed L-tryptophan synthesis from D-serine.
Reaction pathway of tryptophanase-catalyzed L-tryptophan synthesis from D-serine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-05-24)
Akihiko Shimada, Haruka Ozaki, Takeshi Saito, Noriko Fujii
ABSTRACT
Tryptophanase, L-tryptophan indole-lyase with extremely absolute stereospecificity, can change the stereospecificity in concentrated diammonium hydrogenphosphate solution. While tryptophanase is not inert to D-serine in the absence of diammonium hydrogenphosphate, it can undergo L-tryptophan synthesis from D-serine along with indole in the presence of it. It has been well known that tryptophanase synthesizes L-tryptophan from L-serine through a β-substitution mechanism of the ping-pong type. However, a metabolic pathway of L-tryptophan synthesis from D-serine has remained unclear. The present study aims to elucidate it. Diammonium hydrogenphosphate plays a role in the emergence of catalytic activity on D-serine. The salt gives tryptophanase a small conformational change, which makes it possible to catalyze D-serine. Tryptophanase-bound D-serine produces L-tryptophan synthesis by β-replacement reaction via the enzyme-bound aminoacrylate intermediate. Our result will be valuable in studying the origin of homochirality.
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Apotryptophanase from Escherichia coli, soluble powder, 75-150 units/mg solid
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%