MilliporeSigma
  • Heme destruction, the main molecular event during the peroxide-mediated inactivation of chloroperoxidase from Caldariomyces fumago.
Heme destruction, the main molecular event during the peroxide-mediated inactivation of chloroperoxidase from Caldariomyces fumago.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2010-09-14)
Marcela Ayala, Cesar V Batista, Rafael Vazquez-Duhalt
ABSTRACT
Heme peroxidases are subject to a mechanism-based oxidative inactivation. During the catalytic cycle, the heme group is activated to form highly oxidizing species, which may extract electrons from the protein itself. In this work, we analyze changes in residues prone to oxidation owing to their low redox potential during the peroxide-mediated inactivation of chloroperoxidase from Caldariomyces fumago under peroxidasic catalytic conditions. Surprisingly, we found only minor changes in the amino acid content of the fully inactivated enzyme. Our results show that tyrosine residues are not oxidized, whereas all tryptophan residues are partially oxidized in the inactive protein. The data suggest that the main process leading to enzyme inactivation is heme destruction. The molecular characterization of the peroxide-mediated inactivation process could provide specific targets for the protein engineering of this versatile peroxidase.
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, aqueous suspension, brown, >10,000 U/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, ≥3,000 units/mL