MilliporeSigma
  • Engineering Klebsiella oxytoca for efficient 2, 3-butanediol production through insertional inactivation of acetaldehyde dehydrogenase gene.
Engineering Klebsiella oxytoca for efficient 2, 3-butanediol production through insertional inactivation of acetaldehyde dehydrogenase gene.
Applied microbiology and biotechnology (2009-09-17)
Xiao-Jun Ji, He Huang, Jian-Guo Zhu, Lu-Jing Ren, Zhi-Kui Nie, Jun Du, Shuang Li
ABSTRACT
Ethanol was a major byproduct of 2,3-butanediol (2,3-BD) fermentation by Klebsiella oxytoca ME-UD-3. In order to achieve a high efficiency of 2,3-BD production, K. oxytoca mutants deficient in ethanol formation were successfully constructed by replace the aldA gene coding for aldehyde dehydrogenase with a tetracycline resistance cassette. The results suggested that inactivation of aldA led to a significantly improved 2,3-BD production. The carbon flux to 2,3-BD was enhanced by eliminating the byproducing ethanol and at the same time reducing the accumulation of another byproduct acetoin. At last, by fed-batch culturing of the mutant, the final 2,3-BD titer up to 130 g/l with the productivity of 1.63 g/l.h and the 2,3-BD yield relative to glucose of 0.48 g/g was obtained.
Sigma-Aldrich
Aldehyde Dehydrogenase, potassium-activated from baker′s yeast (S. cerevisiae), lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Aldehyde Dehydrogenase, potassium-activated from yeast, lyophilized powder, ≥10 units/mg protein