MilliporeSigma
  • Chitosan-SiO2-multiwall carbon nanotubes nanocomposite: a novel matrix for the immobilization of creatine amidinohydrolase.
Chitosan-SiO2-multiwall carbon nanotubes nanocomposite: a novel matrix for the immobilization of creatine amidinohydrolase.
International journal of biological macromolecules (2009-05-12)
Ashutosh Tiwari, Sanjay R Dhakate
ABSTRACT
A matrix made up of chitosan-SiO(2)-multiwall carbon nanotubes (CHIT-SiO(2)-MWCNTs) nanocomposite was fabricated to investigate the immobilization of creatine amidinohydrolase (CAH). CAH enzyme was covalently immobilized with the CHIT-SiO(2)-MWCNTs matrix using glutaraldehyde as a linker. The resulting CAH/CHIT-SiO(2)-MWCNTs biomatrix was characterized with Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy (SEM) and cyclic voltammetry (CV) taking CHIT-SiO(2)-MWNTs as a reference. The influence of various parameters on CAH enzyme activity within the matrix was investigated including pH, temperature, and time. The Michaelis-Menten constant and apparent activities for the CAH enzyme were calculated to be 0.58 mM and 83.16 mg/cm(2), respectively; indicating CHIT-SiO(2)-MWCNTs nanocomposite matrix has a high affinity to immobilize CAH enzyme.
Sigma-Aldrich
Creatinase from microorganisms, lyophilized powder, ≥4 units/mg solid
Sigma-Aldrich
Creatinase from Pseudomonas sp., recombinant, expressed in E. coli, lyophilized powder, 10-15 units/mg protein