MilliporeSigma
  • Cloning and nucleotide sequence of the Vibrio proteolyticus aminopeptidase gene.
Cloning and nucleotide sequence of the Vibrio proteolyticus aminopeptidase gene.
Biochimica et biophysica acta (1992-07-15)
G Van Heeke, S Denslow, J R Watkins, K J Wilson, F W Wagner
ABSTRACT
The gene encoding the Vibrio proteolyticus aminopeptidase was cloned and sequenced and its amino acid sequence was deduced. The gene encodes a 54 kDa protein, larger than the previously reported size of 30 kDa for the purified aminopeptidase. Sequence alignments revealed a 43-45% homology with two other Vibrio sp. extracellular proteinases.
Sigma-Aldrich
Aminopeptidase His-tagged from Vibrio proteolyticus
Sigma-Aldrich
Aminopeptidase from Aeromonas proteolytica, lyophilized powder, 50-150 units/mg protein