MilliporeSigma
  • Cross-linked aggregates of (R)-oxynitrilase: a stable, recyclable biocatalyst for enantioselective hydrocyanation.
Cross-linked aggregates of (R)-oxynitrilase: a stable, recyclable biocatalyst for enantioselective hydrocyanation.
Organic letters (2005-01-14)
Luuk M van Langen, Rhoderick P Selassa, Fred van Rantwijk, Roger A Sheldon
ABSTRACT
[Reaction: see text] The (R)-oxynitrilase from almonds was immobilized as a cross-linked enzyme aggregate (CLEA) via precipitation with 1,2-dimethoxyethane and subsequent cross-linking using glutaraldehyde. The resulting preparation was a highly effective hydrocyanation catalyst under microaqueous conditions, which suppress the nonenzymatic background reaction. The beneficial effect of these latter conditions on the hydrocyanation of slow-reacting aldehydes is demonstrated. The oxynitrilase CLEA was recycled 10 times without loss of activity.
Product Description
Supelco
1,2-Dimethoxyethane, analytical standard
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
1,2-Dimethoxyethane, suitable for HPLC, 99.9%, inhibitor-free