MilliporeSigma
  • Antioxidative effects of glycosyl-ascorbic acids synthesized by maltogenic amylase to reduce lipid oxidation and volatiles production in cooked chicken meat.
Antioxidative effects of glycosyl-ascorbic acids synthesized by maltogenic amylase to reduce lipid oxidation and volatiles production in cooked chicken meat.
Bioscience, biotechnology, and biochemistry (2004-01-28)
Soo-Bok Lee, Ki-chang Nam, Sung-Joon Lee, Jong-Ho Lee, Kuniyo Inouye, Kwan-Hwa Park
ABSTRACT
Glycosylated ascorbic acids were synthesized by using the transglycosylation activity of Bacillus stearothermophilus maltogenic amylase with maltotriose to show effective antioxidative activity with enhanced oxidative stability. The modified ascorbic acids comprised mono- and di-glycosyl transfer products with an alpha-(1,6)-glycosidic linkage. The antioxidative effects of the glycosyl derivatives of ascorbic acid on the lipid oxidation of cooked chicken breast meat patties were compared, and the synergistic effect when combined with alpha-tocopherol was determined in terms of thiobarbituric acid-reactive substances (TBARS) and volatiles production during storage. The results indicate that the glycosylated ascorbic acids had very effective antioxidative activity in preventing lipid oxidation, and were better in their synergistic effect in comparison to authentic ascorbic acid, with maltosyl-ascorbic acid being the most effective. Volatiles production was highly correlated with the TBARS values in the lipid oxidation of cooked meat. The antioxidative effect preventing the production of volatiles was particularly strong on pentanal, fairly strong on propanal and butanal, and not at all on ethanal. Propanal, pentanal, and the total volatiles thus provided a good representation of the lipid oxidation status of cooked chicken meat.
Sigma-Aldrich
Butyraldehyde, ≥96.0%
Sigma-Aldrich
Butyraldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Butyraldehyde, natural, FG
Sigma-Aldrich
Valeraldehyde, 97%
Sigma-Aldrich
Valeraldehyde, ≥97%, FG
Sigma-Aldrich
Butyraldehyde, ≥99.0%, dry
Sigma-Aldrich
Amylase, Maltogenic from Bacillus sp.