MilliporeSigma
  • Crystallization and preliminary X-ray analysis of substrate complexes of leucine dehydrogenase from Thermoactinomyces intermedius.
Crystallization and preliminary X-ray analysis of substrate complexes of leucine dehydrogenase from Thermoactinomyces intermedius.
Acta crystallographica. Section D, Biological crystallography (2002-05-31)
Tatyana A Muranova, Sergey N Ruzheinikov, Svetlana E Sedelnikova, Patrick J Baker, Alessandra Pasquo, Andrey Galkin, Nobuyoshi Esaki, Toshihisa Ohshima, Kenji Soda, David W Rice
ABSTRACT
Leucine dehydrogenase is an octameric enzyme which belongs to the superfamily of amino-acid dehydrogenases and catalyses the reversible oxidative deamination of leucine to 2-ketoisocaproate, with the corresponding reduction of the cofactor NAD(+). Catalysis by this enzyme is thought to involve a large-scale motion of the enzyme's two domains between an 'open' and 'closed' form, with the latter representing a conformation of the enzyme in which the partners involved in the hydride-transfer reaction are appropriately positioned for catalysis. Whilst a structure for the open form of the enzyme has been determined, the nature of the closed form has yet to be observed. In order to trap a closed form, crystals of the complexes of leucine dehydrogenase from Thermoactinomyces intermedius with 2-ketoisocaproate and with 2-ketoisocaproate and NAD(+) have been obtained by the hanging-drop vapour-diffusion method using PEG 4000 as a precipitant. The crystals of the binary complex with 2-ketoisocaproate belong to space group P2(1)2(1)2(1), with approximate unit-cell parameters a = 106, b = 118, c = 320 A and an octamer in the asymmetric unit, corresponding to a V(M) of 3.1 A(3) Da(-1). The crystals of the non-productive ternary complex belong to space group P6(1) or P6(5), with approximate unit-cell parameters a = b = 117, c = 502 A and an octamer in the asymmetric unit, corresponding to a V(M) of 3.0 A(3) Da(-1). These crystals diffract X-rays on a synchrotron-radiation source to at least 2.8 and 3.3 A resolution, respectively, and are suitable for a full structure determination.
Sigma-Aldrich
L-Leucine Dehydrogenase from Bacillus cereus, lyophilized powder, ≥60 units/mg protein