MilliporeSigma
  • The extracellular regions of PSMA and the transferrin receptor contain an aminopeptidase domain: implications for drug design.
The extracellular regions of PSMA and the transferrin receptor contain an aminopeptidase domain: implications for drug design.
Protein science : a publication of the Protein Society (1999-12-14)
D Mahadevan, J W Saldanha
ABSTRACT
The Aeromonas proteolytica aminopeptidase (AMP), Pseudomonas sp. (RS-16) carboxypeptidase G2 (CPG2), and Streptomyces griseus aminopeptidase (SGAP) are zinc dependent proteolytic enzymes with cocatalytic zinc ion centers and a conserved aminopeptidase fold. A BLAST search with the sequence of the solved AMP structure indicated that a similar domain could be found in prostate-specific membrane antigen (PSMA) and the transferrin receptor (TfR). When the PSMA or TfR sequence was input into the THREADER program, the top structural matches were SGAP and AMP confirming that these are structurally conserved domains. Optimal sequence alignment of PSMA and TfR using the known three-dimensional structures of AMP, CPG2, and SGAP shows that the critical amino acids involved in forming the catalytic pocket are conserved in PSMA but absent in the TfR. The specificity pocket in AMP is formed from four aromatic side chains and the equivalent region in CPG2/PSMA has a changed sequence pattern. Since CPG2 and PSMA are folate hydrolases, the changed specificity pocket leaves space to accommodate the large pteroate moiety of folic acid. In contrast, no enzyme function has been ascribed to the TfR.
Sigma-Aldrich
Aminopeptidase from Aeromonas proteolytica, lyophilized powder, 50-150 units/mg protein
Sigma-Aldrich
Aminopeptidase His-tagged from Vibrio proteolyticus