MilliporeSigma
3'-Phosphatase activity in T4 polynucleotide kinase.
Biochemistry (1977-11-15)
V Cameron, O C Uhlenbeck
PMID199248
ABSTRACT
The purification of T4 polynucleotide kinase results in the copurification of an activity which will specifically remove the 3'-terminal phosphate from a variety of deoxyribonucleotides and ribonucleotides in the absence of ATP. This phosphatase activity requires magnesium, has a pH optiumum of 6.0, and is more active with deoxyribonucleotides than ribonucleotides. T4 polynucleotide kinase and the 3'-phosphatase activity copurify by gradient elution column chromatography on DEAE-cellulose, phosphocellulose, and hydroxylapatite. The two activities are included in and comigrate on Sephadex G-200. Polyacrylamide gel electrophoresis at PH 9.2 results in conigration of the two activities together with the major protein band. The two activities respond in parallel to heat inactivation at 35 degrees C and ATP, a substrate for the kinase only, protects both activities from heat inactivation. It is therefore suggested that the two activities are functions of the same protein molecule.
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution