MilliporeSigma
  • A simple and rapid method for the selection of oligodeoxynucleotide-directed mutants.
A simple and rapid method for the selection of oligodeoxynucleotide-directed mutants.
Gene (1988-05-15)
M A Vandeyar, M P Weiner, C J Hutton, C A Batt
ABSTRACT
We describe an in vitro selection procedure for oligodeoxynucleotide-directed mutagenesis, which produces mutants at frequencies of greater than 90%, facilitating the identification of mutants directly by nucleotide sequencing. The method is based on the selective methylation of the mutant strand by the incorporation of 5-methyl-dCTP. Restriction endonuclease digestion of the resulting hemimethylated DNA with MspI results in the nicking of only the nonmethylated-parental strand. The parental strand is removed by treatment with exonuclease III. The mutants are recovered by transformation of a mcrAB strain of Escherichia coli with the nascent strand.