MilliporeSigma
  • Single-step method of RNA isolation by acid guanidinium thiocyanate-phenol-chloroform extraction.
Single-step method of RNA isolation by acid guanidinium thiocyanate-phenol-chloroform extraction.
Analytical biochemistry (1987-04-01)
P Chomczynski, N Sacchi
ABSTRACT
A new method of total RNA isolation by a single extraction with an acid guanidinium thiocyanate-phenol-chloroform mixture is described. The method provides a pure preparation of undegraded RNA in high yield and can be completed within 4 h. It is particularly useful for processing large numbers of samples and for isolation of RNA from minute quantities of cells or tissue samples.
Sigma-Aldrich
RNA isolation kit, BioUltra
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Phenol:Chloroform:Iso-amyl alcohol (125:24:1)