MilliporeSigma
  • 2-Aminoethyl methylphosphonate, a potent and rapidly acting antagonist of GABA(A)-ρ1 receptors.
2-Aminoethyl methylphosphonate, a potent and rapidly acting antagonist of GABA(A)-ρ1 receptors.
Molecular pharmacology (2011-08-04)
An Xie, Jun Yan, Lan Yue, Feng Feng, Fozia Mir, Heba Abdel-Halim, Mary Chebib, Guy C Le Breton, Robert F Standaert, Haohua Qian, David R Pepperberg
ABSTRACT
2-Aminoethyl methylphosphonate (2-AEMP), an analog of GABA, has been found to exhibit antagonist activity at GABA(A)-ρ1 (also known as ρ1 GABA(C)) receptors. The present study was undertaken to elucidate 2-AEMP's action and to test the activities of 2-AEMP analogs. Whole-cell patch-clamp techniques were used to record membrane currents in neuroblastoma cells stably transfected with human GABA(A)-ρ1 receptors. The action of 2-AEMP was compared with that of 1,2,5,6-tetrahydropyridin-4-yl methylphosphinic acid (TPMPA), a commonly used GABA(A)-ρ1 antagonist. With 10 μM GABA, 2-AEMP's IC(50) (18 μM) differed by less than 2.5-fold from that of TPMPA (7 μM), and results obtained were consistent with a primarily competitive mode of inhibition by 2-AEMP. Terminating the presentation of 2-AEMP or TPMPA in the presence of GABA produced a release from inhibition. However, the rate of inhibition release upon the termination of 2-AEMP considerably exceeded that determined with termination of TPMPA. Moreover, when presented at concentrations near their respective IC(50) values, the preincubation period associated with 2-AEMP's onset of inhibition was much shorter than that for TPMPA. Analogs of 2-AEMP possessing a benzyl or n-butyl rather than a methyl substituent at the phosphorus atom, as well as analogs bearing a C-methyl substituent on the aminoethyl side chain, exhibited reduced potency relative to 2-AEMP. Of these analogs, only (R)-2-aminopropyl methylphosphonate significantly diminished the response to 10 μM GABA. Structure-activity relationships are discussed in the context of molecular modeling of ligand binding to the antagonist binding site of the GABA(A)-ρ1 receptor.
Sigma-Aldrich
Methylphosphonic acid, 98%