MilliporeSigma
  • Use of a phycoerythrin-conjugated anti-glycophorin A monoclonal antibody as a double label to improve the accuracy of FMH quantification by flow cytometry.
Use of a phycoerythrin-conjugated anti-glycophorin A monoclonal antibody as a double label to improve the accuracy of FMH quantification by flow cytometry.
Transfusion medicine (Oxford, England) (1999-06-03)
P Lloyd-Evans, A R Guest, E B Austin, M L Scott
ABSTRACT
The use of flow cytometry for quantifying fetomaternal haemorrhage is increasing, and has been shown to be more accurate than the Kleihauer-Betke test for evaluating larger bleeds of over 4 mL in volume. Red cells are stained with fluorescently labelled monoclonal anti-D. Cells for analysis are normally gated manually on the basis of forward and side scatter. We investigated whether the use of an antiglycophorin A monoclonal antibody conjugate (red cell specific) in a dual labelling technique would improve the gating of RBC and FMH quantification. Mixes of adult rr and cord R1r RBC were prepared to simulate 1, 0.5, 0.25, 0.12 and 0.06% fetal bleeds. Phycoerythrin-conjugated BRIC 256 (mouse monoclonal antiglycophorin A) was used to label all RBC, and FITC-BRAD-3 monoclonal anti-D was used to determine the proportion of D-positive cells. Results from the dual labelling experiments were compared to those from single labelling of the same mixtures with FITC-BRAD-3 alone, using gated and ungated data. The results showed that single labelling with manual gating gave falsely low FMH estimates. We conclude that use of a fluorescently labelled antiglycophorin A antibody improves the accuracy of the FMH measurement by flow cytometry, as manual subjective gating of RBC excludes a higher proportion of fetal than of adult RBC.
Sigma-Aldrich
FluoroTag FITC Conjugation Kit