MilliporeSigma
  • High-resolution genome-wide mapping of AHR and ARNT binding sites by ChIP-Seq.
High-resolution genome-wide mapping of AHR and ARNT binding sites by ChIP-Seq.
Toxicological sciences : an official journal of the Society of Toxicology (2012-08-21)
Raymond Lo, Jason Matthews
ABSTRACT
The aryl hydrocarbon receptor (AHR) and AHR nuclear translocator (ARNT) activated complex regulates genes in response to the environmental contaminant 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). AHR has also emerged as a potential therapeutic target for the treatment of human diseases and different cancers, including breast cancer. To better understand AHR and ARNT signaling in breast cancer cells, we used chromatin immunoprecipitation linked to high-throughput sequencing to identify AHR- and ARNT-binding sites across the genome in TCDD-treated MCF-7 cells. We identified 2594 AHR-bound, 1352 ARNT-bound, and 882 AHR/ARNT cobound regions. No significant differences in the genomic distribution of AHR and ARNT were observed. Approximately 60% of the cobound regions contained at least one core an aryl hydrocarbon response element (AHRE), 5'-GCGTG-3'. AHR/ARNT peak density was the highest within 1 kb of transcription start sites (TSS); however, a number of AHR/ARNT cobound regions were located as far as 100 kb from TSS. De novo motif discovery identified a symmetrical variation of the AHRE (5'-GTGCGTG-3'), as well as FOXA1 and SP1 binding motifs. Microarray analysis identified 104 TCDD-responsive genes where 98 genes were upregulated by TCDD. Of the 104 regulated genes, 69 (66.3%) were associated with an AHR- or ARNT-bound region within 100 kb of their TSS. Overall our study identified AHR/ARNT cobound regions across the genome, revealed the importance but not absolute requirement for an AHRE in AHR/ARNT interactions with DNA, and identified a modified AHRE motif, thereby increasing our understanding of AHR/ARNT signaling pathway.
Sigma-Aldrich
SeqPlex DNA Amplification Kit, For use with high throughput sequencing technologies, Whole Genome Amplification kit designed to facilitate Next Gen Sequencing.
Pricing and availability is not currently available.