MilliporeSigma
  • Re-expression of CEACAM1 long cytoplasmic domain isoform is associated with invasion and migration of colorectal cancer.
Re-expression of CEACAM1 long cytoplasmic domain isoform is associated with invasion and migration of colorectal cancer.
International journal of cancer (2011-03-18)
Junji Ieda, Shozo Yokoyama, Koichi Tamura, Katsunari Takifuji, Tsukasa Hotta, Kenji Matsuda, Yoshimasa Oku, Toru Nasu, Shigehisa Kiriyama, Naoyuki Yamamoto, Yasushi Nakamura, John E Shively, Hiroki Yamaue
ABSTRACT
Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is known to be downregulated at the transcriptional level in adenoma and carcinoma. Recent reports have shown that CEACAM1 is overexpressed at protein level in colorectal cancer and correlated with clinical stage. The reason why colorectal cancer cells re-expressed CEACAM1 remains unclear. The aim of our study was to clarify the implication of CEACAM1 re-expression in colorectal cancer. Immunohistochemical analyses were conducted with CEACAM1 long (CEACAM1-L) or short (CEACAM1-S) cytoplasmic domain-specific antibodies on clinical samples from 164 patients with colorectal cancer. The risk factors for metastasis and survival were calculated for clinical implication of CEACAM1 re-expression. Invasion chamber and wound healing assays were performed for the effect of CEACAM1 expression on invasion and migration of colorectal cancer cells. CEACAM1-L and CEACAM1-S stained with greater intensity at the invasion front than at the luminal surface of tumors. Differences between the long and short cytoplasmic isoform expression levels were observed at the invasion front. Multivariate analysis showed that CEACAM1-L dominance was an independent risk factor for lymph node metastasis, hematogenous metastasis and short survival. The Kaplan-Meier evaluation demonstrated that CEACAM1-L dominance was associated with shorter survival time (p < 0.0001). In the invasion chamber and wound healing assays, CEACAM1-L promoted invasion and migration. Re-expression of CEACAM1 is observed at the invasion front of colorectal cancer. CEACAM1-L dominance is associated with metastasis and shorter survival of the patients with colorectal cancer. CEACAM1-L dominance is important for colorectal cancer cells invasion and migration.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block