MilliporeSigma
  • C3'/C4'-Stereochemical Effects of Digitoxigenin α-L-/α-D-Glycoside in Cancer Cytotoxicity.
C3'/C4'-Stereochemical Effects of Digitoxigenin α-L-/α-D-Glycoside in Cancer Cytotoxicity.
ChemMedChem (2012-11-10)
John W Hinds, Sean B McKenna, Ehesan U Sharif, Hua-Yu L Wang, Novruz G Akhmedov, George A O'Doherty
ABSTRACT
Sweet'n low in stereo: A Wharton reaction was employed along with a diastereoselective palladium-catalyzed glycosylation and other post-glycosylation transformations to synthesize digitoxin analogues. Cytotoxic evaluation against a panel of cancer cell lines uncovered the stereochemical and substitutional limits of the C3'/C4'-hydroxy functionality in digitoxin monosaccharide.
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Sigma-Aldrich
Digitoxigenin
Digitoxin, European Pharmacopoeia (EP) Reference Standard