- SNX5 is essential for efficient macropinocytosis and antigen processing in primary macrophages.
SNX5 is essential for efficient macropinocytosis and antigen processing in primary macrophages.
Macropinocytosis mediates the bulk endocytosis of solute molecules, nutrients and antigens. As this endocytic pathway is considered important in functions associated with immune responses, the molecular mechanisms regulating this pathway in immune cells is of particular significance. However, the regulators of macropinocytosis in primary cells remain poorly defined. Members of the sorting nexin (SNX) family have been implicated in macropinosome biogenesis in cultured cells and here we have analyzed the role of two SNX family members, SNX1 and its binding partner SNX5, in macropinocytosis of mouse primary macrophages. We show that endogenous SNX1 and SNX5 are localised to newly-formed macropinosomes in primary mouse macrophages and, moreover, demonstrate that SNX5 plays an essential role in macropinosome biogenesis. Depletion of SNX5 in bone marrow-derived macrophages dramatically decreased both the number and size of macropinosomes. Depletion of SNX5 also resulted in dramatic reduction in uptake and processing of soluble ovalbumin in macrophages, indicating that the majority of antigen uptake and delivery to late endosomes is via macropinocytosis. By contrast, the absence of SNX1 had no effect on endogenous SNX5 localisation and macropinosome biogenesis using macrophages from SNX1 knockout mice. Therefore, SNX5 can function independently of SNX1 and is a modulator of macropinocytosis that influences the uptake and processing of soluble antigen in primary mouse macrophages.