MilliporeSigma
  • Comparative study of the photoprotolytic reactions of D-luciferin and oxyluciferin.
Comparative study of the photoprotolytic reactions of D-luciferin and oxyluciferin.
The journal of physical chemistry. A (2012-06-16)
Yuval Erez, Itay Presiado, Rinat Gepshtein, Luís Pinto da Silva, Joaquim C G Esteves da Silva, Dan Huppert
ABSTRACT
Optical steady-state and time-resolved spectroscopic methods were used to study the photoprotolytic reaction of oxyluciferin, the active bioluminescence chromophore of the firefly's luciferase-catalyzed reaction. We found that like D-luciferin, the substrate of the firefly bioluminescence reaction, oxyluciferin is a photoacid with pK(a)* value of ∼0.5, whereas the excited-state proton transfer (ESPT) rate coefficient is 2.2 × 10(10) s(-1), which is somewhat slower than that of D-luciferin. The kinetic isotope effect (KIE) on the fluorescence decay of oxyluciferin is 2.5 ± 0.1, the same value as that of D-luciferin. Both chromophores undergo fluorescence quenching in solutions with a pH value below 3.
Sigma-Aldrich
D-Luciferin potassium salt, ≥98.0% (HPLC)