MilliporeSigma
  • Assembly of digitoxin by gold(I)-catalyzed glycosidation of glycosyl o-alkynylbenzoates.
Assembly of digitoxin by gold(I)-catalyzed glycosidation of glycosyl o-alkynylbenzoates.
The Journal of organic chemistry (2011-11-08)
Yuyong Ma, Zhongzhen Li, Hefang Shi, Jian Zhang, Biao Yu
ABSTRACT
Digitoxin, a clinically important cardiac trisaccharide, was assembled efficiently from digitoxigenin and 3,4-di-O-tert-butyldiphenylsilyl-d-digitoxosyl o-cyclopropylethynylbenzoate in 9 steps and 52% overall yield via alternate glycosylation and protecting group manipulation. The present synthesis showcases the advantage of the gold(I)-catalyzed glycosylation protocol in the synthesis of glycoconjugates containing acid-labile 2-deoxysugar linkages.
Sigma-Aldrich
Digitoxigenin
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Digitoxin, European Pharmacopoeia (EP) Reference Standard