MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The tumor inhibitor and antiangiogenic agent withaferin A targets the intermediate filament protein vimentin.
The tumor inhibitor and antiangiogenic agent withaferin A targets the intermediate filament protein vimentin.
Chemistry & biology (2007-06-23)
Paola Bargagna-Mohan, Adel Hamza, Yang-eon Kim, Yik Khuan Abby Ho, Nirit Mor-Vaknin, Nicole Wendschlag, Junjun Liu, Robert M Evans, David M Markovitz, Chang-Guo Zhan, Kyung Bo Kim, Royce Mohan
ABSTRACT
The natural product withaferin A (WFA) exhibits antitumor and antiangiogenesis activity in vivo, which results from this drug's potent growth inhibitory activities. Here, we show that WFA binds to the intermediate filament (IF) protein, vimentin, by covalently modifying its cysteine residue, which is present in the highly conserved alpha-helical coiled coil 2B domain. WFA induces vimentin filaments to aggregate in vitro, an activity manifested in vivo as punctate cytoplasmic aggregates that colocalize vimentin and F-actin. WFA's potent dominant-negative effect on F-actin requires vimentin expression and induces apoptosis. Finally, we show that WFA-induced inhibition of capillary growth in a mouse model of corneal neovascularization is compromised in vimentin-deficient mice. These findings identify WFA as a chemical genetic probe of IF functions, and illuminate a potential molecular target for withanolide-based therapeutics for treating angioproliferative and malignant diseases.
Sigma-Aldrich
Withaferin A, ≥95% (HPLC)