MilliporeSigma
  • A simple and practical agglutination assay for human leucocyte antigen-B27 typing.
A simple and practical agglutination assay for human leucocyte antigen-B27 typing.
Vox sanguinis (2006-06-08)
O Meyer, Z Abdallah, C Schönemann, F Hugo, A Agaylan, H Kiesewetter, A Salama
ABSTRACT
The human leucocyte antigen (HLA) B27 is the most frequently typed single antigen that is associated with diseases. Here, we describe a simple and rapid particle agglutination assay (PaGIA) for HLA-B27 typing. Superparamagnetic particles were coated with a monoclonal antibody to HLA-B27 and subsequently used for testing. Anticoagulated whole-blood samples were obtained from healthy blood donors (n = 194) with known HLA patterns and from patients (n = 51) who had been typed positive for HLA-B27 by flow cytometry. The particles agglutinated only after incubation with HLA-27-positive blood samples, using the ID-microtyping system. Positive reactions were clearly distinguishable from negative reactions in all samples tested. Flow cytometric HLA-B27 typing revealed an indeterminate result in one patient. The new HLA-B27 PaGIA is suitable for rapid typing of HLA-B27. The assay is simple and easy to perform, and can be implemented in any routine laboratory.
Sigma-Aldrich
Streptavidin particles, magnetic, ≥2 U/mg