MilliporeSigma
  • Phase behavior of lipid monolayers containing DPPC and cholesterol analogs.
Phase behavior of lipid monolayers containing DPPC and cholesterol analogs.
Biophysical journal (2006-02-08)
Benjamin L Stottrup, Sarah L Keller
ABSTRACT
We investigate the miscibility phase behavior of lipid monolayers containing a wide variety of sterols. Six of the sterols satisfy a definition from an earlier study of "membrane-active sterols" in bilayers (cholesterol, epicholesterol, lathosterol, dihydrocholesterol, ergosterol, and desmosterol), and six do not (25-hydroxycholesterol, lanosterol, androstenolone, coprostanol, cholestane, and cholestenone). We find that monolayers containing dipalmitoyl phosphatidylcholine mixed with membrane-active sterols generally produce phase diagrams containing two distinct regions of immiscible liquid phases, whereas those with membrane-inactive sterols generally do not. This observation establishes a correlation between lipid monolayers and bilayers. It also demonstrates that the ability to form two regions of immiscibility in monolayers is not one of the biophysical attributes that explains cholesterol's predominance in animal cell membranes. Furthermore, we find unusual phase behavior for dipalmitoyl phosphatidylcholine monolayers containing 25-hydroxycholesterol, which produce both an upper and a lower miscibility transition. The lower transition correlates with a sharp change of slope in the pressure-area isotherm.
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Pricing and availability is not currently available.