MilliporeSigma
  • Cloning and sequencing of the cDNA for human monocyte chemotactic and activating factor (MCAF).
Cloning and sequencing of the cDNA for human monocyte chemotactic and activating factor (MCAF).
Biochemical and biophysical research communications (1989-02-28)
Y Furutani, H Nomura, M Notake, Y Oyamada, T Fukui, M Yamada, C G Larsen, J J Oppenheim, K Matsushima
ABSTRACT
cDNA clones having a nucleotide sequence encoding a human monocyte chemotactic and activating factor (MCAF) were isolated and sequenced. The amino acid sequence deduced from the nucleotide sequence reveals the primary structure of the MCAF precursor to be composed of a putative signal peptide sequence of 23 amino acid residues and a mature MCAF sequence of 76 amino acid residues. The amino acid sequence of MCAF showed 25-55% homology with other members of an inducible cytokine family, including macrophage inflammatory protein and some putative polypeptide mediators known as JE, LD78, RANTES and TCA-3. This suggests that MCAF is a member of family of factors involved in immune and inflammatory responses.