MilliporeSigma
  • Selective in vitro growth of T lymphocytes from normal human bone marrows.
Selective in vitro growth of T lymphocytes from normal human bone marrows.
Science (New York, N.Y.) (1976-09-10)
D A Morgan, F W Ruscetti, R Gallo
PMID181845
ABSTRACT
Selective growth of T lymphocytes occurred when unfractionated normal human bone marrow cells were cultured with conditioned medium obtained from phytohemagglutinin-stimulated normal human lymphocytes (Ly-CM). Cultures of up to 90 percent T cells have been maintained for more than 9 months. The T cells exhibited a strict growth dependence upon Ly-CM and were consistently negative for Epstein-Barr viral information.