MilliporeSigma
  • CyrA, a matricellular protein that modulates cell motility in Dictyostelium discoideum.
CyrA, a matricellular protein that modulates cell motility in Dictyostelium discoideum.
Matrix biology : journal of the International Society for Matrix Biology (2012-03-07)
Robert J Huber, Andres Suarez, Danton H O'Day
ABSTRACT
CyrA, an extracellular matrix (slime sheath), calmodulin (CaM)-binding protein in Dictyostelium discoideum, possesses four tandem EGF-like repeats in its C-terminus and is proteolytically cleaved during asexual development. A previous study reported the expression and localization of CyrA cleavage products CyrA-C45 and CyrA-C40. In this study, an N-terminal antibody was produced that detected the full-length 63kDa protein (CyrA-C63). Western blot analyses showed that the intracellular expression of CyrA-C63 peaked between 12 and 16h of development, consistent with the time that cells are developing into a motile, multicellular slug. CyrA immunolocalization and CyrA-GFP showed that the protein localized to the endoplasmic reticulum, particularly its perinuclear component. CyrA-C63 secretion began shortly after the onset of starvation peaking between 8 and 16h of development. A pharmacological analysis showed that CyrA-C63 secretion was dependent on intracellular Ca(2+) release and active CaM, PI3K, and PLA2. CyrA-C63 bound to CaM both intra- and extracellularly and both proteins were detected in the slime sheath deposited by migrating slugs. In keeping with its purported function, CyrA-GFP over-expression enhanced cAMP-mediated chemotaxis and CyrA-C45 was detected in vinculin B (VinB)-GFP immunoprecipitates, thus providing a link between the increase in chemotaxis and a specific cytoskeletal component. Finally, DdEGFL1-FITC was detected on the membranes of cells capped with concanavalin A suggesting that a receptor exists for this peptide sequence. Together with previous studies, the data presented here suggests that CyrA is a bona fide matricellular protein in D. discoideum.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Sepharose conjugate, buffered aqueous suspension