MilliporeSigma
  • Enhanced exercise capacity in mice with severe heart failure treated with an allosteric effector of hemoglobin, myo-inositol trispyrophosphate.
Enhanced exercise capacity in mice with severe heart failure treated with an allosteric effector of hemoglobin, myo-inositol trispyrophosphate.
Proceedings of the National Academy of Sciences of the United States of America (2009-02-11)
Andreia Biolo, Ruth Greferath, Deborah A Siwik, Fuzhong Qin, Eugene Valsky, Konstantina C Fylaktakidou, Srinivasu Pothukanuri, Carolina D Duarte, Richard P Schwarz, Jean-Marie Lehn, Claude Nicolau, Wilson S Colucci
ABSTRACT
A major determinant of maximal exercise capacity is the delivery of oxygen to exercising muscles. myo-Inositol trispyrophosphate (ITPP) is a recently identified membrane-permeant molecule that causes allosteric regulation of Hb oxygen binding affinity. In normal mice, i.p. administration of ITPP (0.5-3 g/kg) caused a dose-related increase in the oxygen tension at which Hb is 50% saturated (p50), with a maximal increase of 31%. In parallel experiments, ITPP caused a dose-related increase in maximal exercise capacity, with a maximal increase of 57 +/- 13% (P = 0.002). In transgenic mice with severe heart failure caused by cardiac-specific overexpression of G alpha q, i.p. ITPP increased exercise capacity, with a maximal increase of 63 +/- 7% (P = 0.005). Oral administration of ITPP in drinking water increased Hb p50 and maximal exercise capacity (+34 +/- 10%; P < 0.002) in normal and failing mice. Consistent with increased tissue oxygen availability, ITPP decreased hypoxia inducible factor-1alpha mRNA expression in myocardium. It had no effect on myocardial contractility in isolated mouse cardiac myocytes and did not affect arterial blood pressure in vivo in mice. Thus, ITPP decreases the oxygen binding affinity of Hb, increases tissue oxygen delivery, and increases maximal exercise capacity in normal mice and mice with severe heart failure. ITPP is thus an attractive candidate for the therapy of patients with reduced exercise capacity caused by heart failure.
Product Description
Supelco
Amberchrom 50WX8, 200-400 mesh, hydrogen form
Pricing and availability is not currently available.
Sigma-Aldrich
AMBERLITE HPR1100, Na ion exchange resin, strongly acidic, 20-50 mesh
Pricing and availability is not currently available.