MilliporeSigma
Omi, a recessive mutation on chromosome 10, is a novel allele of Ostm1.
Mammalian genome : official journal of the International Mammalian Genome Society (2012-11-20)
Erika A Bosman, Jeanne Estabel, Ozama Ismail, Christine Podrini, Jacqueline K White, Karen P Steel
ABSTRACT
Large-scale N-ethyl-N-nitrosourea (ENU) mutagenesis has provided many rodent models for human disease. Here we describe the initial characterization and mapping of a recessive mutation that leads to degeneration of the incisors, failure of molars to erupt, a grey coat colour, and mild osteopetrosis. We mapped the omi mutation to chromosome 10 between D10Mit214 and D10Mit194. The Ostm1 gene is a likely candidate gene in this region and the grey-lethal allele, Ostm1 ( gl ), and omi mutations fail to complement each other. We show that om/om mice have reduced levels of Ostm1 protein. To date we have not been able to identify the causative mutation. We propose that omi is a novel hypomorphic mutation affecting Ostm1 expression, potentially in a regulatory element.
Sigma-Aldrich
N-Nitroso-N-ethylurea, ISOPAC®
Sigma-Aldrich
N-Nitroso-N-ethylurea, Bulk package