MilliporeSigma
  • Preparation of stable Pd nanocubes and their use in biological labeling.
Preparation of stable Pd nanocubes and their use in biological labeling.
Colloids and surfaces. B, Biointerfaces (2012-07-10)
Miroslav Slouf, Martin Hruby, Zulfiya Bakaeva, Helena Vlkova, Jana Nebesarova, Anatoly A Philimonenko, Pavel Hozak
ABSTRACT
Stable Pd nanocubes (PdNC) with the average size ~15 nm were prepared by the controlled reduction of sodium tetrachloropalladate with ascorbic acid in water, in the presence of polyvinylpyrrolidone and potassium bromide. Morphology of the particles was characterized by transmission electron microscopy (TEM) and their stability in the colloidal solution was verified by dynamic light scattering (DLS). It has been demonstrated that the Pd nanocubes can be distinguished from commercial Au nanospheres in a standard TEM microscope by means of automated image analysis. In the next step, the PdNC were successfully conjugated to immunoglobulin proteins and used for the detection of a specific protein (nucleophosmin) on ultrathin sections of HeLa cells. Our experiments showed that PdNC can be used for multiple immunolabeling in combination with commercial Au nanospheres.
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%