MilliporeSigma
Lactoferrin restrains allergen-induced pleurisy in mice.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2012-07-20)
Michał Zimecki, Jolanta Artym, Maja Kocięba, Katarzyna Kaleta-Kuratewicz, Marian L Kruzel
ABSTRACT
The aim of this study was to assess the utility of lactoferrin (LF), a natural immunomodulator, to restrain allergen-induced pleurisy in mice. BALB/c female mice, 8- to 10-week old, weighing 24 g on average, were used. Mice were immunized intraperitoneally with 50 μg of ovalbumin (OVA) and the pleurisy was elicited 14 days later by intrapleural injection of 12.5 μg of OVA. LF was given 24 and 3 h before elicitation of the allergic reaction. The cytokine levels in the pleural exudates were measured by immunoassays. The blood and pleural exudates smears were stained with Giemsa and May-Grünwald reagents and reviewed histologically. Lung sections were stained with eosin and hematoxylin for histological evaluation. Lactoferrin significantly decreased manifestation of pleurisy induced by OVA in a sensitized mouse model. In particular, the percentages of eosinophils in blood and pleural exudates were strongly diminished. The histological analysis of lungs revealed that LF diminished the development of pathological lesions, such as pulmonary edema, diffuse alveolar hemorrhage and hemosiderosis, which were found in the lungs after injection of the eliciting dose of OVA. LF also decreased the level of IL-5 secreted into the pleural fluid. This is a first demonstration that LF significantly decreases antigen-specific pleurisy in a sensitized mouse model.
Sigma-Aldrich
May-Grünwald solution, for the staining of blood constituents
Sigma-Aldrich
Lactoferrin from human milk, ≥85% (SDS-PAGE), lyophilized powder