MilliporeSigma
  • Phenolic compounds and their anti-oxidative properties and protein kinase inhibition from the Chinese mangrove plant Laguncularia racemosa.
Phenolic compounds and their anti-oxidative properties and protein kinase inhibition from the Chinese mangrove plant Laguncularia racemosa.
Phytochemistry (2009-12-22)
Cui Shi, Min-Juan Xu, Mirko Bayer, Zhi-Wei Deng, Michael H G Kubbutat, Wim Wätjen, Wim Waejen, Peter Proksch, Wen-Han Lin
ABSTRACT
Phenolic compounds, named integracin D (1), (7'R, 8'S, 8S)-8-hydroxyisoguaiacin (3), (2R, 3R) pinobanksin-3-caffeoylate (5) and threo-8S-7-methoxysyringylglycerol (6), respectively, were isolated from the Chinese mangrove plant Laguncularia racemosa (L) Gaertn. f. (Combretaceae), together with 23 known phenolic metabolites. Their structures were elucidated on the basis of extensive spectroscopic analyses including that of IR, UV, MS, CD, 1D and 2D NMR spectra as well as by comparison with literature data. Compound 5 showed significant anti-oxidative activity in the DPPH and TEAC free-radical-scavenging assays, while several of the phenolic compounds were tested for protein kinase inhibitory activity in an assay involving 24 different human tumor related protein kinases. Compounds 5, 7, and 23 showed potential inhibition with IC(50) values between 2.2 and 3.6microg/mL toward individual kinases. The ellagic acid derivatives were tested for insecticidal activity.
Sigma-Aldrich
Pinobanksin, ≥95% (HPLC)