MilliporeSigma
  • The genotoxic effect of potassium metabisulfite using chromosome aberration, sister chromatid exchange, micronucleus tests in human lymphocytes and chromosome aberration test in bone marrow cells of rats.
The genotoxic effect of potassium metabisulfite using chromosome aberration, sister chromatid exchange, micronucleus tests in human lymphocytes and chromosome aberration test in bone marrow cells of rats.
Environmental and molecular mutagenesis (2008-03-14)
Ayse Yavuz-Kocaman, Eyyup Rencuzogullari, Hasan Basri Ila, Mehmet Topaktas
ABSTRACT
Potassium metabisulfite (PMB) is used as an antimicrobial substance in many kinds of foods. In the present study, the effects of PMB on chromosome aberrations (CAs), sister chromatid exchanges (SCEs), and micronucleus (MN) formation in human lymphocytes and as well as its effect on CAs in bone marrow cells of rats were investigated. The human lymphocytes were treated with 25, 50, 100, and 200 microg/ml of PMB for 24 and 48 hr. PMB was also intraperitoneally (ip) injected to the rats as a single dose of 150, 300, and 600 mg/kg body weight (b.w.) for 12 and 24 hr before sacrifice. PMB induced abnormalities such as structural and numerical (total) CAs, SCEs, and MN formations in a dose dependent manner in the lymphocytes of the 24- and 48-hr treatment periods. In addition, PMB showed a cytotoxic effect by decreasing the replication index (RI), mitotic index (MI) and nuclear division index (NDI) in a dose dependent manner in human lymphocytes. The compound induced CA as well and decreased the MI in bone marrow cells of rats. It might be concluded that PMB had a high genotoxic and cytotoxic risk.
Sigma-Aldrich
Potassium disulfite, BioUltra, ≥97.0% (RT)
Sigma-Aldrich
Potassium disulfite, ≥98%
Sigma-Aldrich
Potassium disulfite, puriss., meets analytical specification of Ph.Eur., BP, NF, FCC, E224, 95.0-101.0% (iodometric), powder, 51.8-57.6% sulfur dioxide