MilliporeSigma
New lignan metabolites in rat urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-01-25)
Annika I Smeds, Niina M Saarinen, Patrik C Eklund, Rainer E Sjöholm, Sari I Mäkelä
ABSTRACT
Ten potential lignan metabolites were quantified in rat urine extracts using liquid chromatography-tandem mass spectrometry. The rats were orally administered with the plant lignans 7-hydroxymatairesinol, matairesinol, lariciresinol or secoisolariciresinol, or with the mammalian lignan enterolactone. The samples were enzymatically hydrolysed and solid-phase extracted before analysis. Of the analysed compounds, only trace amounts of 7-oxoenterolactone could be detected in the urine extracts before administration, but after administration of any of the lignans, the excretion of 7-oxoenterolactone increased and monodemethylated matairesinol and 4,4'-dihydroxyenterolactone could be detected. In addition, other novel lignan metabolites were detected, i.e., 7-oxomatairesinol, alpha-conidendrin, and alpha- and beta-conidendric acid.