MilliporeSigma
  • Lipid peroxidation inhibitory activity of some constituents isolated from the stem bark of Eucalyptus globulus.
Lipid peroxidation inhibitory activity of some constituents isolated from the stem bark of Eucalyptus globulus.
Archives of pharmacal research (2000-06-03)
B S Yun, I K Lee, J P Kim, S H Chung, G S Shim, I D Yoo
ABSTRACT
Twelve compounds with lipid peroxidation inhibitory activity were isolated from the stem bark of E. globulus. Their structures were assigned as a new aromatic monoterpene (1) and eleven known compounds, pinoresinol (2), vomifoliol (3), 3,4,5-trimethoxyphenol 1-O-beta-D-(6'-O-galloyl)glucopyranoside (4), methyl gallate (5), rhamnazin (6), rhamnetin (7), eriodictyol (8), quercetin (9), taxifolin (10), engelitin (11), and catechin (12) on the basis of UV, mass, and NMR spectroscopic analyses. These compounds except vomifoliol significantly inhibited lipid peroxidation in rat liver microsome.
Sigma-Aldrich
Rhamnazin, ≥99% (HPLC)