MilliporeSigma
  • A comparison of two procedures for labelling the surface of the hydatid disease organism, Echinococcus granulosus, with 125I.
A comparison of two procedures for labelling the surface of the hydatid disease organism, Echinococcus granulosus, with 125I.
Journal of helminthology (1987-03-01)
D P McManus, D J McLaren, N W Clark, R M Parkhouse
ABSTRACT
Living, intact protoscoleces of the British horse and sheep strains of Echinococcus granulosus were subjected to surface radioiodination procedures using 125I and Iodogen and 125I-Bolton Hunter reagent. Subsequent combined electron microscopy and autoradiography revealed specific surface membrane labelling with the Iodogen procedure, but significant tegumental labelling with the Bolton-Hunter reagent. The two parasite strains yielded different profiles of electrophoretically separated labelled proteins; the Iodogen method, not surprisingly, resulted in a less complex pattern of labelled polypeptides than the Bolton and Hunter reagent.
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester, suitable for fluorescence, BioReagent, ≥97.0% (C)