MilliporeSigma
  • Nonenzymatic displacement of chlorine and formation of free radicals upon the reaction of glutathione with PCB quinones.
Nonenzymatic displacement of chlorine and formation of free radicals upon the reaction of glutathione with PCB quinones.
Proceedings of the National Academy of Sciences of the United States of America (2009-06-06)
Yang Song, Brett A Wagner, Jordan R Witmer, Hans-Joachim Lehmler, Garry R Buettner
ABSTRACT
The reactions of glutathione (GSH) with polychlorinated biphenyl (PCB) quinones having different degrees of chlorination on the quinone ring were examined. EPR spectroscopy and MS revealed 2 types of reactions yielding different products: (i) a nonenzymatic, nucleophilic displacement of chlorine on the quinone ring yielding a glutathiylated conjugated quinone and (ii) Michael addition of GSH to the quinone, a 2-electron reduction, yielding a glutathiylated conjugated hydroquinone. The pK(a) of parent hydroquinone decreased by 1 unit as the degree of chlorination increased. This resulted in a corresponding increase in the oxidizability of these chlorinated hydroquinones. The reaction with oxygen appears to be first-order each in ionized hydroquinone and dioxygen, yielding hydrogen peroxide stoichiometrically. The generation of semiquinone radicals, superoxide, and hydroxyl radicals was observed by EPR; however, the mechanisms and yields vary depending on the degree of the chlorination of hydroquinone/quinone and the presence or absence of GSH. Our discovery that chlorinated quinones undergo a rapid, nonenzymatic dechlorination upon reaction with GSH opens a different view on mechanisms of metabolism and the toxicity of this class of compounds.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, suitable for suitability for complexometry, ≥99.0%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)