MilliporeSigma
  • Induction of human T-cell tolerance to pig xenoantigens via thymus transplantation in mice with an established human immune system.
Induction of human T-cell tolerance to pig xenoantigens via thymus transplantation in mice with an established human immune system.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2009-05-23)
K Habiro, M Sykes, Y-G Yang
ABSTRACT
Thymus xenotransplantation has been shown to induce tolerance to porcine xenografts in mice and to permit survival of alpha1,3Gal-transferase knockout porcine kidney xenografts for months in nonhuman primates. We evaluated the ability of porcine thymus xenotransplantation to induce human T-cell tolerance using a humanized mouse (hu-mouse) model, where a human immune system is preestablished by implantation of fetal human thymus tissue under the kidney capsule and intravenous injection of CD34(+) hematopoietic stem/progenitor cells. Human T-cell depletion with an anti-CD2 mAb following surgical removal of human thymic grafts prevented the initial rejection of porcine thymic xenografts in hu-mice. In these hu-mice, porcine thymic grafts were capable of supporting human thymopoiesis and T-cell development, and inducing human T-cell tolerance to porcine xenoantigens. Human T cells from these mice responded strongly to third-party pig, but not to the thymic donor swine leukocyte antigen (SLA)-matched pig stimulators in a mixed lymphocyte reaction (MLR) assay. Anti-pig xenoreactive antibodies declined in these hu-mice, whereas antibody levels increased in nontolerant animals that rejected porcine thymus grafts. These data show that porcine thymic xenotransplantation can induce donor-specific tolerance in immunocompetent hu-mice, supporting this approach for tolerance induction in clinical xenotransplantation.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Human serum (cystatin C), ERM®, certified reference material