MilliporeSigma
  • Transcription factor C/EBPbeta isoform ratio regulates osteoclastogenesis through MafB.
Transcription factor C/EBPbeta isoform ratio regulates osteoclastogenesis through MafB.
The EMBO journal (2009-05-15)
Jeske J Smink, Valérie Bégay, Ton Schoenmaker, Esta Sterneck, Teun J de Vries, Achim Leutz
ABSTRACT
Disequilibrium between bone-forming osteoblasts and bone-resorbing osteoclasts is central to many bone diseases. Here, we show that dysregulated expression of translationally controlled isoforms of CCAAT/enhancer-binding protein beta (C/EBPbeta) differentially affect bone mass. Alternative translation initiation that is controlled by the mammalian target of rapamycin (mTOR) pathway generates long transactivating (LAP(*), LAP) and a short repressive (LIP) isoforms from a single C/EBPbeta transcript. Rapamycin, an inhibitor of mTOR signalling increases the ratio of LAP over LIP and inhibits osteoclastogenesis in wild type (WT) but not in C/EBPbeta null (c/ebpbeta(-/-)) or in LIP knock-in (L/L) osteoclast precursors. C/EBPbeta mutant mouse strains exhibit increased bone resorption and attenuated expression of MafB, a negative regulator of osteoclastogenesis. Ectopic expression of LAP and LIP in monocytes differentially affect the MafB promoter activity, MafB gene expression and dramatically affect osteoclastogenesis. These data show that mTOR regulates osteoclast formation by modulating the C/EBPbeta isoform ratio, which in turn affects osteoclastogenesis by regulating MafB expression.
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission