MilliporeSigma
  • Quantification of sweat gland innervation: a clinical-pathologic correlation.
Quantification of sweat gland innervation: a clinical-pathologic correlation.
Neurology (2009-04-29)
Christopher H Gibbons, Ben M W Illigens, Ningshan Wang, Roy Freeman
ABSTRACT
To evaluate a novel method to quantify the density of nerve fibers innervating sweat glands in healthy control and diabetic subjects, to compare the results to an unbiased stereologic technique, and to identify the relationship to standardized physical examination and patient-reported symptom scores. Thirty diabetic and 64 healthy subjects had skin biopsies performed at the distal leg and distal and proximal thigh. Nerve fibers innervating sweat glands, stained with PGP 9.5, were imaged by light microscopy. Sweat gland nerve fiber density (SGNFD) was quantified by manual morphometry. As a gold standard, three additional subjects had biopsies analyzed by confocal microscopy using unbiased stereologic quantification. Severity of neuropathy was measured by standardized instruments including the Neuropathy Impairment Score in the Lower Limb (NIS-LL) while symptoms were measured by the Michigan Neuropathy Screening Instrument. Manual morphometry increased with unbiased stereology (r = 0.93, p < 0.01). Diabetic subjects had reduced SGNFD compared to controls at the distal leg (p < 0.001), distal thigh (p < 0.01), and proximal thigh (p < 0.05). The SGNFD at the distal leg of diabetic subjects decreased as the NIS-LL worsened (r = -0.89, p < 0.001) and was concordant with symptoms of reduced sweat production (p < 0.01). We describe a novel method to quantify the density of nerve fibers innervating sweat glands. The technique differentiates groups of patients with mild diabetic neuropathy from healthy control subjects and correlates with both physical examination scores and symptoms relevant to sudomotor dysfunction. This method provides a reliable structural measure of sweat gland innervation that complements the investigation of small fiber neuropathies.
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium permanganate, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)