MilliporeSigma
  • Lentiviral-mediated genetic correction of hematopoietic and mesenchymal progenitor cells from Fanconi anemia patients.
Lentiviral-mediated genetic correction of hematopoietic and mesenchymal progenitor cells from Fanconi anemia patients.
Molecular therapy : the journal of the American Society of Gene Therapy (2009-03-12)
Ariana Jacome, Susana Navarro, Paula Río, Rosa M Yañez, Africa González-Murillo, M Luz Lozano, Maria Luisa Lamana, Julian Sevilla, Teresa Olive, Cristina Diaz-Heredia, Isabel Badell, Jesus Estella, Luis Madero, Guillermo Guenechea, José Casado, Jose C Segovia, Juan A Bueren
ABSTRACT
Previous clinical trials based on the genetic correction of purified CD34(+) cells with gamma-retroviral vectors have demonstrated clinical efficacy in different monogenic diseases, including X-linked severe combined immunodeficiency, adenosine deaminase deficient severe combined immunodeficiency and chronic granulomatous disease. Similar protocols, however, failed to engraft Fanconi anemia (FA) patients with genetically corrected cells. In this study, we first aimed to correlate the hematological status of 27 FA patients with CD34(+) cell values determined in their bone marrow (BM). Strikingly, no correlation between these parameters was observed, although good correlations were obtained when numbers of colony-forming cells (CFCs) were considered. Based on these results, and because purified FA CD34(+) cells might have suboptimal repopulating properties, we investigated the possibility of genetically correcting unselected BM samples from FA patients. Our data show that the lentiviral transduction of unselected FA BM cells mediates an efficient phenotypic correction of hematopoietic progenitor cells and also of CD34(-) mesenchymal stromal cells (MSCs), with a reported role in hematopoietic engraftment. Our results suggest that gene therapy protocols appropriate for the treatment of different monogenic diseases may not be adequate for stem cell diseases like FA. We propose a new approach for the gene therapy of FA based on the rapid transduction of unselected hematopoietic grafts with lentiviral vectors (LVs).
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Essentially fatty acid free
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Supelco
Oil Red O, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)