MilliporeSigma
  • Age-specific induction of mutant p53 drives clonal hematopoiesis and acute myeloid leukemia in adult mice.
Age-specific induction of mutant p53 drives clonal hematopoiesis and acute myeloid leukemia in adult mice.
Cell reports. Medicine (2024-05-12)
Rasoul Pourebrahim, Rafael Heinz Montoya, Hiroki Akiyama, Lauren Ostermann, Shayuan Khazaei, Muharrem Muftuoglu, Natalia Baran, Ran Zhao, Tom Lesluyes, Bin Liu, Joseph D Khoury, Mihai Gagea, Peter Van Loo, Michael Andreeff
ABSTRACT
The investigation of the mechanisms behind p53 mutations in acute myeloid leukemia (AML) has been limited by the lack of suitable mouse models, which historically have resulted in lymphoma rather than leukemia. This study introduces two new AML mouse models. One model induces mutant p53 and Mdm2 haploinsufficiency in early development, showing the role of Mdm2 in myeloid-biased hematopoiesis and AML predisposition, independent of p53. The second model mimics clonal hematopoiesis by inducing mutant p53 in adult hematopoietic stem cells, demonstrating that the timing of p53 mutation determines AML vs. lymphoma development. In this context, age-related changes in hematopoietic stem cells (HSCs) collaborate with mutant p53 to predispose toward myeloid transformation rather than lymphoma development. Our study unveils new insights into the cooperative impact of HSC age, Trp53 mutations, and Mdm2 haploinsufficiency on clonal hematopoiesis and the development of myeloid malignancies.
Sigma-Aldrich
Monoclonal Anti-MDM2 antibody produced in mouse, clone SMP14, ascites fluid