MilliporeSigma
  • Phosphorylation of ATM by Cdk5 mediates DNA damage signalling and regulates neuronal death.
Phosphorylation of ATM by Cdk5 mediates DNA damage signalling and regulates neuronal death.
Nature cell biology (2009-01-20)
Bo Tian, Qian Yang, Zixu Mao
ABSTRACT
The phosphatidylinositol-3-kinase-like kinase ATM (ataxia-telangiectasia mutated) has a central role in coordinating DNA damage responses, including cell-cycle checkpoint control, DNA repair and apoptosis. Mutations of ATM cause a spectrum of defects ranging from neurodegeneration to cancer predisposition. However, the mechanism by which DNA damage activates ATM is poorly understood. Here we show that Cdk5 (cyclin-dependent kinase 5), activated by DNA damage, directly phosphorylates ATM at Ser 794 in post-mitotic neurons. Phosphorylation at Ser 794 precedes, and is required for, ATM autophosphorylation at Ser 1981, and activates ATM kinase activity. The Cdk5-ATM signal regulates phosphorylation and function of the ATM targets p53 and H2AX. Interruption of the Cdk5-ATM pathway attenuates DNA-damage-induced neuronal cell cycle re-entry and expression of the p53 targets PUMA and Bax, protecting neurons from death. Thus, activation of Cdk5 by DNA damage serves as a critical signal to initiate the ATM response and regulate ATM-dependent cellular processes.
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture