MilliporeSigma
  • An algorithm-based technique for counting mitochondria in cells using immunohistochemical staining of formalin-fixed and paraffin-embedded sections.
An algorithm-based technique for counting mitochondria in cells using immunohistochemical staining of formalin-fixed and paraffin-embedded sections.
Journal of cancer research and clinical oncology (2024-04-03)
Mai Sakashita, Noriko Motoi, Gaku Yamamoto, Emi Gambe, Masanori Suzuki, Yukihiro Yoshida, Shun-Ichi Watanabe, Yutaka Takazawa, Kazunori Aoki, Atsushi Ochiai, Shingo Sakashita
ABSTRACT
Visualizing mitochondria in cancer cells from human pathological specimens may improve our understanding of cancer biology. However, using immunohistochemistry to evaluate mitochondria remains difficult because almost all cells contain mitochondria and the number of mitochondria per cell may have important effects on mitochondrial function. Herein, we established an objective system (Mito-score) for evaluating mitochondria using machine-based processing of hue, saturation, and value color spaces. The Mito-score was defined as the number of COX4 (mitochondrial inner membrane) immunohistochemistry-positive pixels divided by the number of nuclei per cell. The system was validated using four lung cancer cell lines, normal tissues, and lung cancer tissues (199 cases). The Mito-score correlated with MitoTracker, a fluorescent dye used to selectively label and visualize mitochondria within cells under a microscope (R2 = 0.68) and with the number of mitochondria counted using electron microscopy (R2 = 0.79). Histologically, the Mito-score of small cell carcinoma (57.25) was significantly lower than that of adenocarcinoma (147.5, p < 0.0001), squamous cell carcinoma (120.6, p = 0.0004), and large cell neuroendocrine carcinoma (111.8, p = 0.002). The Mito-score method enables the analysis of the mitochondrial status of human formalin-fixed paraffin-embedded specimens and may provide insights into the metabolic status of cancer.
Sigma-Aldrich
Anti-COX4I1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-VDAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution